Gene Transcripts

Gene Symbol
OPN1MW
Entrez Gene ID
2652
Gene Name
opsin 1 (cone pigments), medium-wave-sensitive
Gene Aliases
CBBM, CBD, COD5, GCP, GOP, OPN1MW1
Chromosome Location
Chr. X: 154182596 - 154196861 on Build GRCh38
UniGene ID
Hs.247787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000513.2 NP_000504.1 4 788
GenBank mRNA BC140938.1 - 4 788
BC143790.1 - 4 769 9052304
BC171754.1 - 4 769 9053255
BC171755.1 - 4 769 9053256

Gene Symbol
OPN1MW2
Entrez Gene ID
728458
Gene Name
opsin 1 (cone pigments), medium-wave-sensitive 2
Gene Aliases
GOP
Chromosome Location
Chr. X: 154219734 - 154233286 on Build GRCh38
UniGene ID
Hs.571751
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048181.2 NP_001041646.1 4 788

Gene Symbol
OPN1MW3
Entrez Gene ID
101060233
Gene Name
opsin 1 (cone pigments), medium-wave-sensitive 3
Gene Aliases
Chromosome Location
Chr. X: 154257620 - 154271068 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003960089.4 XP_003960138.1 4 706

View Full Product Details