Gene Transcripts

Gene Symbol
AFF3
Entrez Gene ID
3899
Gene Name
AF4/FMR2 family member 3
Gene Aliases
LAF4, MLLT2-like
Chromosome Location
Chr. 2: 99545861 - 100142583 on Build GRCh38
UniGene ID
Hs.444414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011511175.2 XP_011509477.1 2 164
XM_011511177.2 XP_011509479.1 2 164

View Full Product Details