Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016312.2 | NP_057396.1 | 8 | 955 | |
GenBank mRNA | AB029309.1 | - | 8 | 937 | |
AF118023.1 | - | 8 | 873 | ||
AK297410.1 | - | 8 | 979 | ||
AK302244.1 | - | 8 | 993 | ||
BC001621.1 | - | 8 | 955 | 3357963 | |
BC016441.2 | - | 1 | 73 | ||
BC023532.2 | - | 8 | 948 | 4122697 | |
BX647347.1 | - | 7 | 699 |