Gene Transcripts

Gene Symbol
C12orf42
Entrez Gene ID
374470
Gene Name
chromosome 12 open reading frame 42
Gene Aliases
Chromosome Location
Chr. 12: 103047669 - 103525258 on Build GRCh38
UniGene ID
Hs.534649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103526.1 10 1809
XM_011538312.2 XP_011536614.1 9 585
XM_017019281.1 XP_016874770.1 9 582
GenBank mRNA BC039352.1 - 10 1767

View Full Product Details