Gene Transcripts
- Gene Symbol
- ANKRD36
- Entrez Gene ID
- 375248
- Gene Name
- ankyrin repeat domain 36
- Gene Aliases
- UNQ2430
- Chromosome Location
- Chr. 2: 97113186 - 97264521 on Build GRCh38
- UniGene ID
- Hs.541894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164315.1 |
NP_001157787.1
|
41, 42 |
2818 |
|
XM_006712514.3 |
XP_006712577.1
|
41, 42 |
3128 |
|
XM_011511130.1 |
XP_011509432.1
|
41, 42 |
3128 |
|
XM_011511131.1 |
XP_011509433.1
|
41, 42 |
3128 |
|
XM_017004009.1 |
XP_016859498.1
|
41, 42 |
3128 |
|
XM_017004010.1 |
XP_016859499.1
|
41, 42 |
3128 |
|
XM_017004011.1 |
XP_016859500.1
|
41, 42 |
3128 |
|
XM_017004012.1 |
XP_016859501.1
|
41, 42 |
3128 |
|
XM_017004013.1 |
XP_016859502.1
|
39, 40 |
3026 |
|
XM_017004014.1 |
XP_016859503.1
|
41, 42 |
3128 |
|
XM_017004015.1 |
XP_016859504.1
|
39, 40 |
3026 |
|
XM_017004016.1 |
XP_016859505.1
|
41, 42 |
3128 |
|
XM_017004017.1 |
XP_016859506.1
|
39, 40 |
3026 |
|
XM_017004018.1 |
XP_016859507.1
|
39, 40 |
3026 |
|
XM_017004019.1 |
XP_016859508.1
|
39, 40 |
3026 |
|
XM_017004020.1 |
XP_016859509.1
|
39, 40 |
3026 |
|
XM_017004021.1 |
XP_016859510.1
|
41, 42 |
3128 |
|
XM_017004022.1 |
XP_016859511.1
|
39, 40 |
3026 |
|
XM_017004023.1 |
XP_016859512.1
|
41, 42 |
3128 |
|
XM_017004024.1 |
XP_016859513.1
|
40, 41 |
2990 |
|
XM_017004025.1 |
XP_016859514.1
|
41, 42 |
3128 |
|
XM_017004026.1 |
XP_016859515.1
|
41, 42 |
3128 |
|
XM_017004027.1 |
XP_016859516.1
|
41, 42 |
3128 |
|
XM_017004028.1 |
XP_016859517.1
|
37, 38 |
2924 |
|
XM_017004029.1 |
XP_016859518.1
|
37, 38 |
2924 |
|
XM_017004031.1 |
XP_016859520.1
|
41, 42 |
3128 |
|
XM_017004032.1 |
XP_016859521.1
|
37, 38 |
2924 |
|
XM_017004033.1 |
XP_016859522.1
|
41, 42 |
3128 |
|
XM_017004034.1 |
XP_016859523.1
|
41, 42 |
3128 |
|
XM_017004035.1 |
XP_016859524.1
|
37, 38 |
2924 |
|
XM_017004036.1 |
XP_016859525.1
|
41, 42 |
3128 |
|
XM_017004037.1 |
XP_016859526.1
|
41, 42 |
3128 |
|
XM_017004038.1 |
XP_016859527.1
|
41, 42 |
3128 |
|
XM_017004039.1 |
XP_016859528.1
|
35, 36 |
2822 |
|
XM_017004041.1 |
XP_016859530.1
|
41, 42 |
3128 |
|
XM_017004042.1 |
XP_016859531.1
|
41, 42 |
3128 |
|
XM_017004043.1 |
XP_016859532.1
|
41, 42 |
3128 |
|
XM_017004045.1 |
XP_016859534.1
|
41, 42 |
3128 |
|
XM_017004046.1 |
XP_016859535.1
|
39, 40 |
2883 |
|
XM_017004047.1 |
XP_016859536.1
|
38, 39 |
2815 |
|
XM_017004048.1 |
XP_016859537.1
|
37, 38 |
2742 |
|
XM_017004049.1 |
XP_016859538.1
|
39, 40 |
2917 |
|
XM_017004050.1 |
XP_016859539.1
|
24, 25 |
1321 |
|
XM_017004051.1 |
XP_016859540.1
|
22, 23 |
1535 |
|
GenBank mRNA |
BC128262.1 |
- |
7, 8 |
905 |
|
DA952980.1 |
- |
4, 5 |
192 |
|
View Full Product Details