Gene Transcripts

Gene Symbol
Adcy6
Entrez Gene ID
11512
Gene Name
adenylate cyclase 6
Gene Aliases
AC6, mKIAA0422
Chromosome Location
Chr. 15: 98589978 - 98610140 on Build GRCm38
UniGene ID
Mm.157091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007405.2 NP_031431.2 19, 20 3202
GenBank mRNA AK122279.1 - 5, 6 1146
AK147516.1 - 19, 20 3202
BC145101.1 - 18, 19 3000
M93422.1 - 18, 19 3134
M96653.1 - 18, 19 3024

View Full Product Details