Gene Transcripts

Gene Symbol
Akap3
Entrez Gene ID
11642
Gene Name
A kinase (PRKA) anchor protein 3
Gene Aliases
Prka3
Chromosome Location
Chr. 6: 126853098 - 126874308 on Build GRCm38
UniGene ID
Mm.87748
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009650.2 NP_033780.2 5 2352
GenBank mRNA AF093406.1 - 5 2278
BC100458.1 - 5 2225 30917596

View Full Product Details