Gene Transcripts
- Gene Symbol
- Araf
- Entrez Gene ID
- 11836
- Gene Name
- v-raf murine sarcoma 3611 viral oncogene homolog
- Gene Aliases
- 1200013E08Rik, A-Raf, AW495444, Araf1
- Chromosome Location
- Chr. X: 20797787 - 20860521 on Build GRCm38
- UniGene ID
- Mm.220946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159645.1 |
NP_001153117.1
|
4, 5 |
438 |
|
NM_009703.2 |
NP_033833.1
|
4, 5 |
438 |
|
GenBank mRNA |
AB158252.1 |
- |
3, 4 |
363 |
|
AK004741.1 |
- |
3, 4 |
1717 |
|
AK010060.1 |
- |
2, 3 |
189 |
|
AK020547.1 |
- |
4, 5 |
404 |
|
AK034861.1 |
- |
4, 5 |
433 |
|
AK039044.1 |
- |
4, 5 |
404 |
|
AK085823.1 |
- |
4, 5 |
404 |
|
AK133797.1 |
- |
3, 4 |
1724 |
|
AK135358.1 |
- |
4, 5 |
404 |
|
AK163567.1 |
- |
4, 5 |
407 |
|
AK169757.1 |
- |
4, 5 |
419 |
|
BB857915.1 |
- |
4, 5 |
438 |
|
BC004757.1 |
- |
4, 5 |
394 |
3154293 |
View Full Product Details