Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009710.4 | NP_033840.2 | 3 | 900 | |
GenBank mRNA | AK003107.1 | - | 3 | 901 | |
AK009284.1 | - | 3 | 229 | ||
AK009902.1 | - | 901 | |||
BC063760.1 | - | 3 | 881 | 30137479 | |
BC132492.1 | - | 3 | 888 | 40130769 | |
BC132496.1 | - | 3 | 888 | 40130773 | |
X95842.1 | - | 2 | 862 |