Gene Transcripts
- Gene Symbol
- Astn1
- Entrez Gene ID
- 11899
- Gene Name
- astrotactin 1
- Gene Aliases
- Astn, GC14, mKIAA0289
- Chromosome Location
- Chr. 1: 158362102 - 158691786 on Build GRCm38
- UniGene ID
- Mm.329586
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001205204.1 |
NP_001192133.1
|
12, 13 |
2362 |
|
NM_007495.4 |
NP_031521.2
|
12, 13 |
2338 |
|
GenBank mRNA |
AK045561.1 |
- |
12, 13 |
2093 |
|
AK138576.1 |
- |
12, 13 |
2107 |
|
AK147450.1 |
- |
12, 13 |
2108 |
|
AK164117.1 |
- |
12, 13 |
2327 |
|
AK165200.1 |
- |
12, 13 |
2091 |
|
BC054545.1 |
- |
9, 10 |
1112 |
|
BC094666.1 |
- |
12, 13 |
2224 |
30545499 |
G49230.1 |
- |
12, 13 |
1948 |
|
U48797.1 |
- |
12, 13 |
1948 |
|
View Full Product Details