Gene Transcripts

Gene Symbol
Atp5k
Entrez Gene ID
11958
Gene Name
ATP synthase, H+ transporting, mitochondrial F1F0 complex, subunit E
Gene Aliases
2610008D24Rik, Atp5i, Lfm1
Chromosome Location
Chr. 5: 108433253 - 108434378 on Build GRCm38
UniGene ID
Mm.136093
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007507.2 NP_031533.2 3 168
GenBank mRNA BC028438.1 - 3 168
S52977.1 - 3 151

View Full Product Details