Gene Transcripts

Gene Symbol
Bche
Entrez Gene ID
12038
Gene Name
butyrylcholinesterase
Gene Aliases
C730038G20Rik
Chromosome Location
Chr. 3: 73635805 - 73708437 on Build GRCm38
UniGene ID
Mm.250719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009738.3 NP_033868.3 2 797
GenBank mRNA AK050337.1 - 2 798
AK052200.1 - 2 626
BC099977.1 - 2 624 6751601
CK636839.1 - 1 520
M99492.1 - 1 525

View Full Product Details