Gene Transcripts
- Gene Symbol
- Bgn
- Entrez Gene ID
- 12111
- Gene Name
- biglycan
- Gene Aliases
- BG, DSPG1, PG-S1, PGI, SLRR1A
- Chromosome Location
- Chr. X: 73483612 - 73495936 on Build GRCm38
- UniGene ID
- Mm.2608
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007542.4 |
NP_031568.2
|
3, 4 |
502 |
|
GenBank mRNA |
AK135496.1 |
- |
3, 4 |
503 |
|
AK155973.1 |
- |
3, 4 |
503 |
|
AK164867.1 |
- |
3, 4 |
503 |
|
AK171870.1 |
- |
3, 4 |
503 |
|
BC005452.1 |
- |
3, 4 |
496 |
3594469 |
BC019502.1 |
- |
3, 4 |
505 |
4237268 |
BC052857.1 |
- |
3, 4 |
487 |
30061850 |
L20276.1 |
- |
3, 4 |
502 |
|
X53928.1 |
- |
|
597 |
|
Y11758.1 |
- |
2, 3 |
333 |
|
View Full Product Details