Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007573.2 | NP_031599.2 | 5, 6 | 774 | |
GenBank mRNA | AF300619.1 | - | 5, 6 | 713 | |
AJ001101.1 | - | 5, 6 | 719 | ||
AK010601.1 | - | 5, 6 | 712 | ||
AK010746.1 | - | 5, 6 | 738 | ||
AK011868.1 | - | 5, 6 | 765 | ||
AK169764.1 | - | 5, 6 | 757 | ||
BC038075.1 | - | 5, 6 | 758 | 3154276 |