Gene Transcripts

Gene Symbol
Camk4
Entrez Gene ID
12326
Gene Name
calcium/calmodulin-dependent protein kinase IV
Gene Aliases
A430110E23Rik, AI666733, CaMKIV, CaMKIV/Gr, D18Bwg0362e
Chromosome Location
Chr. 18: 32934957 - 33195767 on Build GRCm38
UniGene ID
Mm.222329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009793.3 NP_033923.2 2 571
GenBank mRNA AK030219.1 - 2 455
AK043114.1 - 2 281
BC070420.1 - 2 452 5718423
X58995.1 - 2 261

View Full Product Details