Gene Transcripts
- Gene Symbol
- Casp3
- Entrez Gene ID
- 12367
- Gene Name
- caspase 3
- Gene Aliases
- A830040C14Rik, AC-3, Apopain, CASP-3, CC3, CPP-32, CPP32, Caspase-3, Lice, SCA-1, Yama, mldy
- Chromosome Location
- Chr. 8: 46617291 - 46639698 on Build GRCm38
- UniGene ID
- Mm.34405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284409.1 |
NP_001271338.1
|
4, 5 |
534 |
|
NM_009810.3 |
NP_033940.1
|
4, 5 |
529 |
|
GenBank mRNA |
AK080651.1 |
- |
1, 2 |
44 |
|
AK146226.1 |
- |
4, 5 |
535 |
|
BC038825.1 |
- |
4, 5 |
418 |
4482368 |
D86352.1 |
- |
4, 5 |
348 |
|
U19522.1 |
- |
4, 5 |
345 |
|
U49929.1 |
- |
4, 5 |
371 |
|
U63720.1 |
- |
1, 2 |
137 |
|
Y13086.1 |
- |
4, 5 |
375 |
|
View Full Product Details