Gene Transcripts

Gene Symbol
Fas
Entrez Gene ID
14102
Gene Name
Fas (TNF receptor superfamily member 6)
Gene Aliases
AI196731, APO1, APT1, CD95, TNFR6, Tnfrsf6, lpr
Chromosome Location
Chr. 19: 34290659 - 34327775 on Build GRCm38
UniGene ID
Mm.1626
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007987.2 NP_032013.2 4 411
GenBank mRNA AK002590.1 - 4 362
BC061160.1 - 4 384 30302649
DQ846748.1 - 4 355
M83649.1 - 4 404

View Full Product Details