Gene Transcripts

Gene Symbol
Itln1
Entrez Gene ID
16429
Gene Name
intelectin 1 (galactofuranose binding)
Gene Aliases
IntL, Itln, Itln2, Itln5, Itlna, Lfr
Chromosome Location
Chr. 1: 171518123 - 171535325 on Build GRCm38
UniGene ID
Mm.247426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010584.3 NP_034714.1 6, 7 775
GenBank mRNA AB016496.1 - 6, 7 774
AK007447.1 - 6, 7 775
AY157363.1 - 6, 7 764
AY157364.1 - 6, 7 759
AY619693.1 - 6, 7 758
BC145377.1 - 6, 7 721 9053935
BC150760.1 - 6, 7 721 9087671
BC150795.1 - 6, 7 717 9087706

View Full Product Details