Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080798.1 | NP_001074267.1 | 3 | 534 | |
NM_133919.3 | NP_598680.3 | 3 | 545 | ||
GenBank mRNA | AF074266.1 | - | 3 | 378 | |
AK082878.1 | - | 4 | 602 | ||
AK142554.1 | - | 3 | 545 | ||
AK143742.1 | - | 3 | 534 | ||
AY749168.1 | - | 3 | 378 | ||
BC131665.1 | - | 3 | 378 | 40110795 |