Gene Transcripts

Gene Symbol
Myo10
Entrez Gene ID
17909
Gene Name
myosin X
Gene Aliases
AW048724, D15Ertd600e, mKIAA0799
Chromosome Location
Chr. 15: 25622550 - 25813673 on Build GRCm38
UniGene ID
Mm.60590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019472.2 NP_062345.2 25 3735
GenBank mRNA AJ249706.2 - 25 3359
AK122371.1 - 5 1517

View Full Product Details