Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042726.3 | NP_001036191.1 | 1 | 282 | |
NM_021543.4 | NP_067518.2 | 1 | 282 | ||
GenBank mRNA | AF231125.1 | - | 1 | 203 | |
AK048638.1 | - | 1 | 283 | ||
AK049982.1 | - | 1 | 283 | ||
AK140407.1 | - | 1 | 167 | ||
BC052388.1 | - | 1 | 263 | 5718846 | |
BC053008.1 | - | 1 | 270 | 5698009 |