Gene Transcripts

Gene Symbol
Abcb4
Entrez Gene ID
18670
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 4
Gene Aliases
Mdr2, Pgy-2, Pgy2, mdr-2
Chromosome Location
Chr. 5: 8893598 - 8959226 on Build GRCm38
UniGene ID
Mm.297825
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008830.2 NP_032856.2 23 3061
GenBank mRNA AA239419.1 - 2 194
BC150687.1 - 23 2931 9087598
J03398.1 - 23 3061

View Full Product Details