Gene Transcripts

Gene Symbol
Rab12
Entrez Gene ID
19328
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
2900054P15Rik, AW555448, C77700
Chromosome Location
Chr. 17: 66494512 - 66519934 on Build GRCm38
UniGene ID
Mm.248313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024448.2 NP_077768.2 3 403
GenBank mRNA AB232607.1 - 3 303
AK013690.1 - 2 59
AK081084.1 - 1 2501
CK624286.1 - 3 404

View Full Product Details