Gene Transcripts
- Gene Symbol
- Rps17
- Entrez Gene ID
- 20068
- Gene Name
- ribosomal protein S17
- Gene Aliases
- Chromosome Location
- Chr. 7: 81342733 - 81345234 on Build GRCm38
- UniGene ID
- Mm.42767
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009092.3 |
NP_033118.1
|
3, 4 |
287 |
|
GenBank mRNA |
AK007992.1 |
- |
3, 4 |
295 |
|
AK010647.1 |
- |
3, 4 |
289 |
|
AK012781.1 |
- |
3, 4 |
276 |
|
AK158902.1 |
- |
2, 3 |
353 |
|
AK162022.1 |
- |
3, 4 |
281 |
|
AK162335.1 |
- |
3, 4 |
279 |
|
BC081466.1 |
- |
2, 3 |
274 |
6412006 |
BC086900.1 |
- |
3, 4 |
280 |
6760363 |
BC086901.1 |
- |
3, 4 |
280 |
6763239 |
D25213.1 |
- |
2, 3 |
268 |
|
View Full Product Details