Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011348.2 | NP_035478.2 | 16 | 2493 | |
GenBank mRNA | AF034744.1 | - | 16 | 2228 | |
AK129116.1 | - | 11 | 1258 | ||
AK141295.1 | - | 15 | 3216 | ||
AK142340.1 | - | 16 | 2320 | ||
AK159214.1 | - | 16 | 2284 | ||
BC057956.1 | - | 16 | 2391 | 5357516 | |
Z80941.1 | - | 16 | 1859 | ||
Z93947.1 | - | 16 | 2471 | ||
Z93948.1 | - | 16 | 2471 |