Gene Transcripts

Gene Symbol
Sftpc
Entrez Gene ID
20389
Gene Name
surfactant associated protein C
Gene Aliases
Bricd6, SP-C, SP5, SPC, Sftp-2, Sftp2, pro-SpC
Chromosome Location
Chr. 14: 70520941 - 70524081 on Build GRCm38
UniGene ID
Mm.24040
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011359.2 NP_035489.2 3 266
GenBank mRNA BC061137.1 - 3 261
BY773034.1 - 3 267
CJ093163.1 - 3 262

View Full Product Details