Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011459.4 | NP_035589.1 | 7 | 1095 | |
GenBank mRNA | AK032659.1 | - | 7 | 1094 | |
AK036811.1 | - | 7 | 1031 | ||
BC011076.1 | - | 7 | 1075 | ||
BC025121.1 | - | 7 | 1085 | 4487039 | |
U96703.2 | - | 7 | 1086 |