Gene Transcripts

Gene Symbol
Spp1
Entrez Gene ID
20750
Gene Name
secreted phosphoprotein 1
Gene Aliases
2AR, Apl-1, BNSP, BSPI, Bsp, ETA-1, Eta, OP, Opn, Opnl, Ric, Spp-1
Chromosome Location
Chr. 5: 104435111 - 104441053 on Build GRCm38
UniGene ID
Mm.288474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204201.1 NP_001191130.1 7 481
NM_001204202.1 NP_001191131.1 6 478
NM_001204203.1 NP_001191132.1 7 429
NM_001204233.1 NP_001191162.1 5 523
NM_009263.3 NP_033289.2 6 426
GenBank mRNA AF515708.1 - 5 326
AK133555.1 - 6 417
AK145099.1 - 6 420
AK149588.1 - 6 417
AK150819.1 - 6 417
AK150845.1 - 6 417
AK151069.1 - 6 387
AK151562.1 - 6 417
AK153089.1 - 6 416
AK154908.1 - 6 417
AK160540.1 - 6 420
AK165393.1 - 6 417
AK165965.1 - 6 469
AK166395.1 - 6 420
AK171158.1 - 6 417
BC002113.1 - 6 423 3489075
BC014284.1 - 1 85
BC020355.1 - 6 406
BC029991.1 - 6 406
BC057858.1 - 6 399 5294167
BC080720.1 - 6 399 5252869
CT010357.1 - 5 326
J04806.1 - 6 395
S78177.1 - 4 281
X13986.1 - 5 341
X16151.1 - 5 548

View Full Product Details