Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175247.3 | NP_780456.2 | 7 | 2184 | |
GenBank mRNA | AK028202.1 | - | 7 | 2157 | |
AK048530.1 | - | 7 | 2176 | ||
AK173170.1 | - | 7 | 2149 | ||
AY528713.1 | - | 7 | 2090 | ||
BC062116.1 | - | 7 | 2152 | 6817400 |