Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175247.3 | NP_780456.2 | 7 | 999 | |
GenBank mRNA | AK028202.1 | - | 7 | 972 | |
AK048530.1 | - | 7 | 991 | ||
AK173170.1 | - | 7 | 964 | ||
AY528713.1 | - | 7 | 905 | ||
BC062116.1 | - | 7 | 967 | 6817400 | |
M36516.1 | - | 122 |