Gene Transcripts

Gene Symbol
Btd
Entrez Gene ID
26363
Gene Name
biotinidase
Gene Aliases
Chromosome Location
Chr. 14: 31640999 - 31668197 on Build GRCm38
UniGene ID
Mm.282679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025295.4 NP_079571.1 4 598
GenBank mRNA AK005506.1 - 4 591
AK135375.1 - 4 588
BC024051.1 - 4 565 5099802

View Full Product Details