Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102458.1 | NP_001095928.1 | 13 | 2003 | |
NM_018745.5 | NP_061215.1 | 12 | 1843 | ||
GenBank mRNA | AF032128.1 | - | 10 | 1269 | |
AK049680.1 | - | 12 | 1844 | ||
AK088112.1 | - | 12 | 1841 | ||
AK088671.1 | - | 13 | 2000 | ||
AK133914.1 | - | 12 | 1839 | ||
BC019412.1 | - | 12 | 1831 | 5151406 | |
BC021152.1 | - | 12 | 1796 | ||
BC043722.1 | - | 10 | 1349 | 5321733 | |
BC046814.1 | - | 12 | 1895 |