Gene Transcripts

Gene Symbol
Camkk1
Entrez Gene ID
55984
Gene Name
calcium/calmodulin-dependent protein kinase kinase 1, alpha
Gene Aliases
AI846603, CaMKKalpha, Camkk
Chromosome Location
Chr. 11: 73015306 - 73042073 on Build GRCm38
UniGene ID
Mm.9998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018883.2 NP_061371.2 4 601
GenBank mRNA AF117384.1 - 4 556
AK144483.1 - 4 602
BC017529.1 - 4 566 4924656

View Full Product Details