Gene Transcripts

Gene Symbol
Slc2a8
Entrez Gene ID
56017
Gene Name
solute carrier family 2, (facilitated glucose transporter), member 8
Gene Aliases
D2Ertd44e, GLUT8, GlutX1
Chromosome Location
Chr. 2: 32972981 - 32982113 on Build GRCm38
UniGene ID
Mm.305754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019488.4 NP_062361.1 5 668
GenBank mRNA AF232061.2 - 5 625
AJ245936.1 - 5 645
AK081806.1 - 5 685
AK141193.1 - 4 834
AK155792.1 - 5 697
AK170319.1 - 5 681
AY856043.1 - 5 629
AY856044.1 - 5 629
AY856045.1 - 5 629
AY856046.1 - 2 159
BC090993.1 - 5 668 6751763
Y17802.1 - 5 650

View Full Product Details