Gene Transcripts

Gene Symbol
Scube2
Entrez Gene ID
56788
Gene Name
signal peptide, CUB domain, EGF-like 2
Gene Aliases
4932442O19Rik, Cegf1, Cegp1
Chromosome Location
Chr. 7: 109798690 - 109865679 on Build GRCm38
UniGene ID
Mm.40447
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020052.2 NP_064436.2 11 1310
GenBank mRNA AK016571.1 - 5 457
AK142950.1 - 11 1276

View Full Product Details