Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027015.4 | NP_081291.1 | 2 | 130 | |
GenBank mRNA | AA036204.1 | - | 2 | 284 | |
AI851912.1 | - | 2 | 365 | ||
AK014294.1 | - | 2 | 130 | ||
AK084058.1 | - | 2 | 130 | ||
AK088320.1 | - | 2 | 133 | ||
AK166551.1 | - | 2 | 131 | ||
BC027649.1 | - | 2 | 117 | ||
BC037026.1 | - | 2 | 130 | ||
BC048352.1 | - | 2 | 114 | 5008906 | |
BC055693.1 | - | 2 | 120 | 6408041 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190258.1 | NP_001177187.1 | 1 | 129 |