Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021895.2 | NP_068695.1 | 2 | 344 | |
GenBank mRNA | AJ289242.2 | - | 2 | 344 | |
AK145322.1 | - | 2 | 397 | ||
AK150044.1 | - | 2 | 346 | ||
BC013616.1 | - | 2 | 353 | 3157024 | |
BC027032.1 | - | 2 | 295 | ||
BC087554.1 | - | 2 | 297 | 5351470 | |
DQ303411.1 | - | 2 | 247 | ||
U41415.1 | - | 1 | 31 |