Gene Transcripts

Gene Symbol
Rmnd5b
Entrez Gene ID
66089
Gene Name
required for meiotic nuclear division 5 homolog B (S. cerevisiae)
Gene Aliases
0610039K22Rik, Gid2
Chromosome Location
Chr. 11: 51623673 - 51635896 on Build GRCm38
UniGene ID
Mm.288183
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025346.1 NP_079622.1 7 1148
GenBank mRNA AK005344.1 - 4 527
AK075599.1 - 7 1148
BC016075.1 - 7 1002 4485143

View Full Product Details