Gene Transcripts

Gene Symbol
Asb17
Entrez Gene ID
66772
Gene Name
ankyrin repeat and SOCS box-containing 17
Gene Aliases
4933436O18Rik
Chromosome Location
Chr. 3: 153844247 - 153853615 on Build GRCm38
UniGene ID
Mm.116960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025758.4 NP_080034.2 2 631
GenBank mRNA AF425641.1 - 2 631
AK017090.1 - 2 640
AK164225.1 - 78
BC049572.1 - 2 636 6773716

View Full Product Details