Gene Transcripts

Gene Symbol
Rnmtl1
Entrez Gene ID
67390
Gene Name
RNA methyltransferase like 1
Gene Aliases
4833420N02Rik, AI255153, HC90
Chromosome Location
Chr. 11: 76243736 - 76250625 on Build GRCm38
UniGene ID
Mm.24886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183263.5 NP_899086.2 3 646
GenBank mRNA AK012200.1 - 3 620
AK014739.1 - 3 612
AK147805.1 - 3 627
BC090975.1 - 3 618 6595681

View Full Product Details