Gene Transcripts

Gene Symbol
Pnpla8
Entrez Gene ID
67452
Gene Name
patatin-like phospholipase domain containing 8
Gene Aliases
1200006O19Rik, AI467579, Ipla2(gamma)
Chromosome Location
Chr. 12: 44269128 - 44315935 on Build GRCm38
UniGene ID
Mm.54126
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026164.2 NP_080440.2 3 1291
GenBank mRNA AB044139.1 - 3 1170
AK004621.1 - 3 1258
AK084928.1 - 958
AK145776.1 - 3 1201
AK163211.1 - 3 1259
AK168475.1 - 3 1254
BC127056.1 - 2 1151 40109753

View Full Product Details