Gene Transcripts

Gene Symbol
Zfand4
Entrez Gene ID
67492
Gene Name
zinc finger, AN1-type domain 4
Gene Aliases
Anubl1
Chromosome Location
Chr. 6: 116264209 - 116330379 on Build GRCm38
UniGene ID
Mm.272772
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290338.1 NP_001277267.1 6 1886
NM_001290339.1 NP_001277268.1 4 1586
GenBank mRNA AK006693.1 - 1 493
AK012639.1 - 4 1585
AK089032.1 - 6 1883
CX223310.1 - 1 18

View Full Product Details