Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271696.1 | NP_001258625.1 | 14 | 1979 | |
NM_001271697.1 | NP_001258626.1 | 13 | 1859 | ||
NM_001271698.1 | NP_001258627.1 | 13 | 1901 | ||
NM_001271699.1 | NP_001258628.1 | 13 | 1862 | ||
NM_004299.4 | NP_004290.2 | 14 | 1982 | ||
GenBank mRNA | AF038950.1 | - | 14 | 1919 | |
AF078777.1 | - | 14 | 1899 | ||
AF133659.1 | - | 14 | 1911 | ||
AK001418.1 | - | 14 | 1939 | ||
AK294657.1 | - | 13 | 1843 | ||
AK307414.1 | - | 13 | 1782 | ||
BC006323.2 | - | 14 | 1915 | 4138236 | |
BT009918.1 | - | 14 | 1914 | ||
BX537833.1 | - | 13 | 1799 | ||
CN411218.1 | - | 1 | 27 | ||
KU177872.1 | - | 14 | 1914 | ||
KU177873.1 | - | 12 | 1556 | ||
KU177874.1 | - | 4 | 480 | ||
KU177875.1 | - | 5 | 440 | ||
U66679.1 | - | 5 | 632 |