Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr. 17: 44759037 - 44781846 on Build GRCh38
UniGene ID
Hs.6088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318933.1 NP_001305862.1 19 1605
NM_002390.5 NP_002381.2 19 1609
XM_005257373.3 XP_005257430.1 19 1751
XM_017024647.1 XP_016880136.1 18 1523
GenBank mRNA AB009675.1 - 19 1588
AK296512.1 - 19 1686
CN418442.1 - 3 222
D17390.1 - 16 1318

View Full Product Details