Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033874.2 | NP_001029046.2 | 4, 5 | 371 | |
GenBank mRNA | AK004439.1 | - | 4, 5 | 369 | |
BC100481.1 | - | 4, 5 | 368 | 30917849 | |
BC109352.2 | - | 4, 5 | 358 | 40057006 |