Gene Transcripts
- Gene Symbol
- MFNG
- Entrez Gene ID
- 4242
- Gene Name
- MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
- Gene Aliases
- Chromosome Location
- Chr. 22: 37469063 - 37486440 on Build GRCh38
- UniGene ID
- Hs.517603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166343.1 |
NP_001159815.1
|
3, 4 |
733 |
|
NM_002405.3 |
NP_002396.2
|
4, 5 |
828 |
|
NR_029413.1 |
|
3, 4 |
779 |
|
GenBank mRNA |
AK292123.1 |
- |
4, 5 |
769 |
|
AK297149.1 |
- |
3, 4 |
727 |
|
AK297833.1 |
- |
3, 4 |
712 |
|
AK316555.1 |
- |
3, 4 |
720 |
|
BC094814.1 |
- |
4, 5 |
789 |
5930073 |
BM457814.1 |
- |
4, 5 |
783 |
|
CR456518.1 |
- |
4, 5 |
621 |
|
U94352.1 |
- |
4, 5 |
735 |
|
View Full Product Details