Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159505.1 | NP_001152977.1 | 1 | 415 | |
NM_023517.2 | NP_076006.2 | 1 | 415 | ||
GenBank mRNA | AK009514.1 | - | 1 | 417 | |
AK040667.1 | - | 1 | 193 | ||
AK134028.1 | - | 1 | 121 | ||
AK170351.1 | - | 1 | 422 | ||
BC069900.1 | - | 1 | 374 | 5290965 |