Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002446.3 | NP_002437.2 | 1 | 428 | |
XM_011526981.2 | XP_011525283.1 | 1 | 515 | ||
XM_011526982.2 | XP_011525284.1 | 1 | 514 | ||
GenBank mRNA | X90846.1 | - | 1 | 428 | |
Z48615.1 | - | 1 | 300 |