Gene Transcripts

Gene Symbol
Ttl
Entrez Gene ID
69737
Gene Name
tubulin tyrosine ligase
Gene Aliases
2410003M22Rik, 2700049H19Rik, AI848570
Chromosome Location
Chr. 2: 129065947 - 129096283 on Build GRCm38
UniGene ID
Mm.33909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027192.2 NP_081468.1 5 868
GenBank mRNA AK010374.1 - 3 244
BC018513.1 - 5 871 3597859
BC024414.1 - 5 840 4973810

View Full Product Details