Gene Transcripts

Gene Symbol
Agk
Entrez Gene ID
69923
Gene Name
acylglycerol kinase
Gene Aliases
2610037M15Rik, 6720408I04Rik, AI465370, Mulk
Chromosome Location
Chr. 6: 40325155 - 40397562 on Build GRCm38
UniGene ID
Mm.32840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023538.2 NP_076027.1 2, 3 163
GenBank mRNA AJ401619.1 - 2, 3 136
AK011715.1 - 2, 3 159
AK076224.1 - 1, 2 126
AK089433.1 - 2, 3 134
AK155346.1 - 2, 3 129
AY772469.1 - 1, 2 104
BC019145.1 - 2, 3 157 5051535
BC093525.1 - 2, 3 133 5151866

View Full Product Details